Gene Transcripts

Gene Symbol
MIEF1
Entrez Gene ID
54471
Gene Name
mitochondrial elongation factor 1
Gene Aliases
HSU79252, MID51, SMCR7L, dJ1104E15.3
Chromosome Location
Chr. 22: 39500100 - 39518134 on Build GRCh38
UniGene ID
Hs.728085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304564.1 NP_001291493.1 6 1285
NM_019008.5 NP_061881.2 6 1335
NR_130789.1 6 1249
NR_130790.1 7 1399
XM_011530236.1 XP_011528538.1 8 1403
XM_011530237.1 XP_011528539.1 7 1523
XM_011530238.1 XP_011528540.1 7 1607
XM_017028838.1 XP_016884327.1 7 1501
GenBank mRNA AK290954.1 - 7 1232
AL365515.1 - 6 1212
AL834205.1 - 6 1177
BC002587.2 - 2 477
BC008327.1 - 6 1189 3502711

View Full Product Details