Gene Transcripts

Gene Symbol
TOLLIP
Entrez Gene ID
54472
Gene Name
toll interacting protein
Gene Aliases
IL-1RAcPIP
Chromosome Location
Chr. 11: 1274368 - 1309666 on Build GRCh38
UniGene ID
Hs.368527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318512.1 NP_001305441.1 2 359
NM_001318514.1 NP_001305443.1 3 519
NM_019009.3 NP_061882.2 3 509
GenBank mRNA AB209541.1 - 3 450
AJ242972.1 - 3 455
AK022871.1 - 3 416
AK090850.1 - 2 320
AK127123.1 - 3 472
AK315592.1 - 3 470
AL136835.1 - 3 456
AY730683.1 - 3 345
BC004420.1 - 3 469 3356339
BC012057.1 - 3 434 4647828
BC018272.1 - 3 426 3452585
CR533477.1 - 3 345
DB039994.1 - 3 510

View Full Product Details