Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019069.3 | NP_061942.2 | 1 | 652 | |
GenBank mRNA | AK002149.1 | - | 1 | 380 | |
AK315186.1 | - | 1 | 270 | ||
BC025762.1 | - | 1 | 652 | ||
BC043494.1 | - | 1 | 241 | 6056052 | |
CR457279.1 | - | 1 | 145 |