Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005507.2 | NP_005498.1 | 2 | 490 | |
GenBank mRNA | AK097690.1 | - | 3 | 972 | |
AK303614.1 | - | 2 | 334 | ||
BC011005.2 | - | 2 | 318 | 4079579 | |
BC012265.1 | - | 2 | 323 | 3862540 | |
BC012318.1 | - | 2 | 322 | 3538185 | |
BC018256.2 | - | 2 | 318 | 3448134 | |
BC024975.1 | - | 1 | 25 | ||
BT006846.1 | - | 1 | 256 | ||
D00682.1 | - | 1 | 256 | ||
EU794603.1 | - | 1 | 256 | ||
U21909.1 | - | 1 | 256 | ||
X95404.1 | - | 2 | 307 |