Gene Transcripts

Gene Symbol
MBTD1
Entrez Gene ID
54799
Gene Name
mbt domain containing 1
Gene Aliases
SA49P01
Chromosome Location
Chr. 17: 51177419 - 51260511 on Build GRCh38
UniGene ID
Hs.656803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017643.2 NP_060113.2 10, 11 1400
XM_005257467.4 XP_005257524.1 9, 10 1345
XM_005257468.4 XP_005257525.1 9, 10 1338
XM_011524922.2 XP_011523224.1 9, 10 1347
XM_011524923.2 XP_011523225.1 9, 10 1344
XM_011524924.2 XP_011523226.1 9, 10 1340
XM_011524925.1 XP_011523227.1 10, 11 1461
XM_011524926.2 XP_011523228.1 10, 11 1565
XM_011524928.2 XP_011523230.1 9, 10 1502
XM_011524929.2 XP_011523231.1 10, 11 1416
XM_011524930.1 XP_011523232.1 9, 10 1138
XM_011524931.2 XP_011523233.1 8, 9 972
XM_011524933.2 XP_011523235.1 11, 12 1522
XM_011524934.2 XP_011523236.1 9, 10 1346
XM_011524935.2 XP_011523237.1 9, 10 1339
XM_011524936.1 XP_011523238.1 9, 10 1138
XM_017024782.1 XP_016880271.1 10, 11 1712
GenBank mRNA AK000062.1 - 6, 7 732
AK124061.1 - 9, 10 1138
BC034364.1 - 10, 11 1371
BC101736.1 - 5, 6 661 8069242

View Full Product Details