Gene Transcripts

Gene Symbol
FNBP1L
Entrez Gene ID
54874
Gene Name
formin binding protein 1 like
Gene Aliases
C1orf39, TOCA1
Chromosome Location
Chr. 1: 93448067 - 93569671 on Build GRCh38
UniGene ID
Hs.134060
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024948.2 NP_001020119.1 6 606
NM_001164473.2 NP_001157945.1 6 606
NM_017737.4 NP_060207.2 6 606
XM_011541625.2 XP_011539927.1 6 670
XM_017001533.1 XP_016857022.1 7 801
GenBank mRNA AY514449.1 - 6 455
BC062477.1 - 1 57

View Full Product Details