Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271762.1 | NP_001258691.1 | 3, 4 | 592 | |
NM_001271763.1 | NP_001258692.1 | 4, 5 | 672 | ||
NM_017803.4 | NP_060273.1 | 4, 5 | 672 | ||
GenBank mRNA | AB101210.1 | - | 4, 5 | 336 | |
AK000406.1 | - | 3, 4 | 239 | ||
AK290578.1 | - | 4, 5 | 374 | ||
AK296663.1 | - | 4, 5 | 324 | ||
BC006527.2 | - | 4, 5 | 315 | 2988128 |