Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017831.3 | NP_060301.2 | 2 | 665 | |
XM_011526045.2 | XP_011524347.1 | 2 | 934 | ||
XM_011526046.2 | XP_011524348.1 | 2 | 315 | ||
XM_011526047.2 | XP_011524349.1 | 2 | 944 | ||
GenBank mRNA | AK000463.1 | - | 2 | 665 | |
AK225518.1 | - | 2 | 665 | ||
BC012021.1 | - | 2 | 296 | 4521247 | |
BM469263.1 | - | 2 | 296 | ||
BU500546.1 | - | 2 | 155 |