Gene Transcripts

Gene Symbol
TTC22
Entrez Gene ID
55001
Gene Name
tetratricopeptide repeat domain 22
Gene Aliases
Chromosome Location
Chr. 1: 54779696 - 54802379 on Build GRCh38
UniGene ID
Hs.16230
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114108.1 NP_001107580.1 2, 3 724
NM_017904.3 NP_060374.2 2, 3 724
XM_011541671.2 XP_011539973.1 2, 3 1835
XM_017001582.1 XP_016857071.1 2, 3 1858
GenBank mRNA AK000626.1 - 2, 3 654

View Full Product Details