Gene Transcripts

Gene Symbol
HEATR3
Entrez Gene ID
55027
Gene Name
HEAT repeat containing 3
Gene Aliases
SYO1
Chromosome Location
Chr. 16: 50065970 - 50107268 on Build GRCh38
UniGene ID
Hs.733027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182922.2 NP_891552.1 6 839
XM_005256013.2 XP_005256070.1 5 654
XM_011523188.2 XP_011521490.1 6 729
XM_017023385.1 XP_016878874.1 4 792
XM_017023386.1 XP_016878875.1 4 597
GenBank mRNA AF462442.1 - 5 646
AK000725.1 - 5 646
AK289949.1 - 6 804
BC014415.1 - 816
BC018730.1 - 6 804 4907047
BC033077.1 - 6 705 4301559

View Full Product Details