Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282659.1 | NP_001269588.1 | 18 | 2903 | |
NM_017944.3 | NP_060414.3 | 17 | 2699 | ||
XM_005252997.1 | XP_005253054.1 | 19 | 2963 | ||
XM_011520196.1 | XP_011518498.1 | 11 | 2078 | ||
XM_017017953.1 | XP_016873442.1 | 19 | 3019 | ||
XM_017017954.1 | XP_016873443.1 | 11 | 3226 | ||
GenBank mRNA | AK027362.1 | - | 17 | 2104 | |
AK074076.1 | - | 3 | 391 | ||
AK092290.1 | - | 19 | 2319 | ||
AK127264.1 | - | 18 | 2699 | ||
AL832991.2 | - | 3 | 314 |