Gene Transcripts

Gene Symbol
ATG16L1
Entrez Gene ID
55054
Gene Name
autophagy related 16 like 1
Gene Aliases
APG16L, ATG16A, ATG16L, IBD10, WDR30
Chromosome Location
Chr. 2: 233251571 - 233295674 on Build GRCh38
UniGene ID
Hs.529322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190266.1 NP_001177195.1 6, 7 961
NM_001190267.1 NP_001177196.1 6, 7 959
NM_017974.3 NP_060444.3 6, 7 963
NM_030803.6 NP_110430.5 6, 7 963
NM_198890.2 NP_942593.2 3, 4 531
XM_005246082.2 XP_005246139.1 6, 7 900
XM_005246084.2 XP_005246141.1 3, 4 439
XM_005246086.2 XP_005246143.1 3, 4 439
XM_006712608.2 XP_006712671.1 5, 6 711
GenBank mRNA AK000897.1 - 5, 6 574
AK001690.1 - 6, 7 809
AK024453.1 - 6, 7 833
AK027854.1 - 5, 6 545
AK123876.1 - 3, 4 430
AK308400.1 - 6, 7 863
AY358182.1 - 6, 7 852
AY398617.1 - 6, 7 852
BC071846.1 - 3, 4 369
BC117337.1 - 6, 7 625
BC143900.1 - 6, 7 625
EF079889.1 - 6, 7 706
EF079890.1 - 3, 4 274
EF560734.1 - 6, 7 833
JQ924062.1 - 6, 7 706

View Full Product Details