Gene Transcripts

Gene Symbol
LRRC8D
Entrez Gene ID
55144
Gene Name
leucine rich repeat containing 8 family member D
Gene Aliases
LRRC5
Chromosome Location
Chr. 1: 89821014 - 89936432 on Build GRCh38
UniGene ID
Hs.482087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134479.1 NP_001127951.1 3 1413
NM_018103.4 NP_060573.2 3 1400
XM_011541689.1 XP_011539991.1 4 1484
XM_017001599.1 XP_016857088.1 3 1755
XM_017001600.1 XP_016857089.1 3 1210
XM_017001601.1 XP_016857090.1 3 2101
XM_017001602.1 XP_016857091.1 3 1454
GenBank mRNA AK001332.1 - 1 466
AK092238.1 - 3 1376
AK222787.1 - 1 427
AL831880.1 - 1 872
AL832952.1 - 1 831
BC024159.2 - 3 1367 3958069

View Full Product Details