Gene Transcripts

Gene Symbol
UBR7
Entrez Gene ID
55148
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Gene Aliases
C14orf130
Chromosome Location
Chr. 14: 93207056 - 93229215 on Build GRCh38
UniGene ID
Hs.648806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175748.3 NP_786924.2 9 1332
NR_038150.1 8 1198
GenBank mRNA AK001345.1 - 6 710
AK097477.1 - 9 1317
AK315492.1 - 9 1131
BC015046.1 - 9 1131 3908032
BC017013.1 - 8 976 3883770
BC051819.1 - 9 1094 6171393
BX248249.1 - 9 1091

View Full Product Details