Gene Transcripts

Gene Symbol
RBM38
Entrez Gene ID
55544
Gene Name
RNA binding motif protein 38
Gene Aliases
HSRNASEB, RNPC1, SEB4B, SEB4D, dJ800J21.2
Chromosome Location
Chr. 20: 57391398 - 57409333 on Build GRCh38
UniGene ID
Hs.236361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291780.1 NP_001278709.1 1 284
NM_017495.5 NP_059965.2 1 284
NM_183425.2 NP_906270.1 1 284
XM_005260446.3 XP_005260503.1 1 222
XM_011528885.2 XP_011527187.1 1 216
GenBank mRNA AF432218.1 - 1 280
BC018711.1 - 1 36 4852523
BM549757.1 - 279
DN998561.1 - 1 269
X75314.1 - 1 71

View Full Product Details