Gene Transcripts

Gene Symbol
STAB2
Entrez Gene ID
55576
Gene Name
stabilin 2
Gene Aliases
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1
Chromosome Location
Chr. 12: 103587250 - 103766724 on Build GRCh38
UniGene ID
Hs.408249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017564.9 NP_060034.9 28 3213
XM_011538537.2 XP_011536839.1 28 3254
XM_011538538.2 XP_011536840.1 28 3254
XM_011538539.2 XP_011536841.1 14 1457
XM_011538541.2 XP_011536843.1 6 564
XM_017019585.1 XP_016875074.1 28 3254
GenBank mRNA AB052958.1 - 28 3209
AJ295695.1 - 28 3213
AK074051.1 - 6 584
AK160380.1 - 27 2926
AY311388.1 - 28 3231

View Full Product Details