Gene Transcripts
- Gene Symbol
- LMBR1L
- Entrez Gene ID
- 55716
- Gene Name
- limb development membrane protein 1 like
- Gene Aliases
- LIMR
- Chromosome Location
- Chr. 12: 49096551 - 49111535 on Build GRCh38
- UniGene ID
- Hs.272838
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300750.1 |
NP_001287679.1
|
14, 15 |
1622 |
|
NM_018113.3 |
NP_060583.2
|
13, 14 |
1420 |
|
XM_011538563.2 |
XP_011536865.1
|
13, 14 |
1574 |
|
XM_011538564.2 |
XP_011536866.1
|
13, 14 |
1947 |
|
XM_011538565.2 |
XP_011536867.1
|
12, 13 |
2196 |
|
XM_011538566.2 |
XP_011536868.1
|
12, 13 |
1170 |
|
XM_017019626.1 |
XP_016875115.1
|
13, 14 |
1822 |
|
XM_017019628.1 |
XP_016875117.1
|
13, 14 |
1409 |
|
XM_017019630.1 |
XP_016875119.1
|
10, 11 |
3382 |
|
GenBank mRNA |
AF260728.1 |
- |
13, 14 |
1379 |
|
AK001356.1 |
- |
14, 15 |
1403 |
|
AL137599.1 |
- |
10, 11 |
813 |
|
AY358572.1 |
- |
13, 14 |
1384 |
|
BC015015.1 |
- |
13, 14 |
1149 |
3910520 |
BC031550.1 |
- |
13, 14 |
1181 |
5194023 |
KU178711.1 |
- |
13, 14 |
1075 |
|
KU178712.1 |
- |
12, 13 |
1058 |
|
KU178713.1 |
- |
12, 13 |
1041 |
|
View Full Product Details