Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018229.3 | NP_060699.3 | 5 | 1589 | |
NR_026895.1 | 4 | 943 | |||
XM_006720196.2 | XP_006720259.1 | 5 | 1218 | ||
XM_011536940.2 | XP_011535242.1 | 6 | 1561 | ||
GenBank mRNA | AK001675.1 | - | 5 | 1434 | |
AK172824.1 | - | 4 | 651 | ||
AK225345.1 | - | 5 | 1436 | ||
AL136685.1 | - | 5 | 1568 |