Gene Transcripts
- Gene Symbol
- CLCN4
- Entrez Gene ID
- 1183
- Gene Name
- chloride voltage-gated channel 4
- Gene Aliases
- CLC4, ClC-4, ClC-4A
- Chromosome Location
- Chr. X: 10156945 - 10237660 on Build GRCh38
- UniGene ID
- Hs.495674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X77197.1 |
- |
11, 12 |
2351 |
|
View Full Product Details