Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr. 9: 33921693 - 34048949 on Build GRCh38
UniGene ID
Hs.493739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282529.1 NP_001269458.1 19 2375
NM_001282530.1 NP_001269459.1 7 918
NM_018449.3 NP_060919.3 23 2689
GenBank mRNA AB040924.1 - 11 1491
AK025357.1 - 4 331
AK025801.1 - 12 1579
AK026088.1 - 15 1857
AK057564.1 - 4 1244
AK074884.1 - 10 1048
AK125935.1 - 6 2173
AK128424.1 - 1 545
AK294952.1 - 23 2607
AK299183.1 - 17 2097
AK302754.1 - 22 2557
AK303493.1 - 19 2313
BC037416.1 - 4 322

View Full Product Details