Gene Transcripts
- Gene Symbol
- UNC45A
- Entrez Gene ID
- 55898
- Gene Name
- unc-45 myosin chaperone A
- Gene Aliases
- GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A
- Chromosome Location
- Chr. 15: 90929980 - 90954093 on Build GRCh38
- UniGene ID
- Hs.389461
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039675.1 |
NP_001034764.1
|
5, 6 |
1008 |
|
NM_001323619.1 |
NP_001310548.1
|
2, 3 |
267 |
|
NM_001323620.1 |
NP_001310549.1
|
2, 3 |
657 |
|
NM_001323621.1 |
NP_001310550.1
|
2, 3 |
612 |
|
NM_018671.4 |
NP_061141.2
|
2, 3 |
657 |
|
XM_005254963.3 |
XP_005255020.1
|
2, 3 |
229 |
|
XM_011521779.2 |
XP_011520081.1
|
5, 6 |
1253 |
|
XM_011521780.2 |
XP_011520082.1
|
1, 2 |
159 |
|
XM_017022407.1 |
XP_016877896.1
|
2, 3 |
229 |
|
GenBank mRNA |
AB014729.1 |
- |
2, 3 |
199 |
|
AB014736.1 |
- |
2, 3 |
241 |
|
AK000905.1 |
- |
2, 3 |
233 |
|
AK125721.1 |
- |
5, 6 |
1008 |
|
AK291622.1 |
- |
2, 3 |
212 |
|
AK293538.1 |
- |
2, 3 |
233 |
|
AK302979.1 |
- |
2, 3 |
274 |
|
BC006214.1 |
- |
2, 3 |
240 |
3051384 |
BC037992.1 |
- |
2, 3 |
240 |
5534906 |
BC045635.1 |
- |
5, 6 |
1008 |
5297091 |
View Full Product Details