Gene Transcripts

Gene Symbol
TEX13B
Entrez Gene ID
56156
Gene Name
testis expressed 13B
Gene Aliases
TGC3B, TSGA5
Chromosome Location
Chr. X: 107980864 - 107982370 on Build GRCh38
UniGene ID
Hs.333130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031273.2 NP_112563.1 2 198
GenBank mRNA AF285598.1 - 2 198
BC150140.1 - 1 337

View Full Product Details