Gene Transcripts
- Gene Symbol
- TMEM9B
- Entrez Gene ID
- 56674
- Gene Name
- TMEM9 domain family member B
- Gene Aliases
- C11orf15
- Chromosome Location
- Chr. 11: 8947201 - 8965011 on Build GRCh38
- UniGene ID
- Hs.501853
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286094.1 |
NP_001273023.1
|
2, 3 |
675 |
|
NM_001286095.1 |
NP_001273024.1
|
3, 4 |
324 |
|
NM_020644.2 |
NP_065695.1
|
3, 4 |
767 |
|
GenBank mRNA |
AB097017.1 |
- |
3, 4 |
392 |
|
AK074677.1 |
- |
3, 4 |
385 |
|
AK300712.1 |
- |
3, 4 |
247 |
|
AK310723.1 |
- |
2, 3 |
306 |
|
AK313065.1 |
- |
3, 4 |
397 |
|
AK316216.1 |
- |
2, 3 |
306 |
|
AY359069.1 |
- |
3, 4 |
410 |
|
BC015884.1 |
- |
2, 3 |
302 |
3909465 |
BC040124.1 |
- |
3, 4 |
449 |
6053804 |
DC314550.1 |
- |
3, 4 |
324 |
|
View Full Product Details