Gene Transcripts

Gene Symbol
CA10
Entrez Gene ID
56934
Gene Name
carbonic anhydrase 10
Gene Aliases
CA-RPX, CARPX, HUCEP-15
Chromosome Location
Chr. 17: 51630313 - 52160017 on Build GRCh38
UniGene ID
Hs.463466
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082533.1 NP_001076002.1 9 2044
NM_001082534.1 NP_001076003.1 9 1609
NM_020178.4 NP_064563.1 8 1918
XM_017024878.1 XP_016880367.1 6 772
GenBank mRNA AB001597.1 - 8 932
AB036836.1 - 9 1834
AF288385.1 - 9 1600
AK294652.1 - 7 1289
AK313001.1 - 8 1494
AL161812.1 - 8 934
AY358509.1 - 8 1212
BC020577.1 - 9 1576 4181377
BC047456.1 - 8 1831 5276935
BC068462.1 - 8 1877 5260321
CR749607.1 - 9 1380

View Full Product Details