Gene Transcripts

Gene Symbol
SDR39U1
Entrez Gene ID
56948
Gene Name
short chain dehydrogenase/reductase family 39U member 1
Gene Aliases
C14orf124, HCDI
Chromosome Location
Chr. 14: 24439766 - 24442905 on Build GRCh38
UniGene ID
Hs.643552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020195.2 NP_064580.2 4 285
XM_011536975.1 XP_011535277.1 4 329
GenBank mRNA AF226050.1 - 3 559
AK123513.1 - 2 727
BC000989.2 - 4 285 2900140
BC008566.1 - 3 457
BI256676.1 - 2 148
DT220226.1 - 4 289

View Full Product Details