Gene Transcripts
- Gene Symbol
- OTUD7B
- Entrez Gene ID
- 56957
- Gene Name
- OTU deubiquitinase 7B
- Gene Aliases
- CEZANNE, ZA20D1
- Chromosome Location
- Chr. 1: 149940330 - 150010766 on Build GRCh38
- UniGene ID
- Hs.98322
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020205.3 |
NP_064590.2
|
9, 10 |
1480 |
|
XM_011509782.1 |
XP_011508084.1
|
9, 10 |
1477 |
|
XM_011509783.2 |
XP_011508085.1
|
9, 10 |
1707 |
|
XM_011509784.2 |
XP_011508086.1
|
9, 10 |
1271 |
|
XM_011509785.2 |
XP_011508087.1
|
9, 10 |
4904 |
|
XM_011509786.1 |
XP_011508088.1
|
9, 10 |
1343 |
|
XM_011509787.1 |
XP_011508089.1
|
9, 10 |
1086 |
|
XM_011509788.1 |
XP_011508090.1
|
8, 9 |
1195 |
|
XM_017001850.1 |
XP_016857339.1
|
9, 10 |
1343 |
|
XM_017001851.1 |
XP_016857340.1
|
8, 9 |
1329 |
|
GenBank mRNA |
AJ293573.1 |
- |
9, 10 |
1323 |
|
AK299790.1 |
- |
9, 10 |
1466 |
|
BC020622.1 |
- |
9, 10 |
1266 |
|
BC072681.1 |
- |
9, 10 |
1417 |
6170513 |
View Full Product Details