Gene Transcripts

Gene Symbol
SLC45A4
Entrez Gene ID
57210
Gene Name
solute carrier family 45 member 4
Gene Aliases
Chromosome Location
Chr. 8: 141207166 - 141308321 on Build GRCh38
UniGene ID
Hs.372492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080431.2 NP_001073900.1 4 1304
NM_001286646.1 NP_001273575.1 4 1549
NM_001286648.1 NP_001273577.1 4 1304
XM_011517169.2 XP_011515471.1 5 1590
XM_011517170.2 XP_011515472.1 6 2395
XM_011517171.2 XP_011515473.1 5 1645
XM_011517172.2 XP_011515474.1 5 2134
XM_011517173.2 XP_011515475.1 5 1590
XM_011517174.2 XP_011515476.1 4 1304
XM_011517176.2 XP_011515478.1 4 1401
XM_011517177.2 XP_011515479.1 4 1842
XM_011517178.2 XP_011515480.1 4 4019
XM_011517179.2 XP_011515481.1 4 1134
XM_017013648.1 XP_016869137.1 5 2669
XM_017013649.1 XP_016869138.1 5 1574
XM_017013650.1 XP_016869139.1 4 2036
GenBank mRNA AB032952.1 - 2 700
AK128212.1 - 4 1304
BC090891.1 - 4 1065
BC157840.1 - 4 1046 9057059
BC171836.1 - 4 1046

View Full Product Details