Gene Transcripts

Gene Symbol
ODF2L
Entrez Gene ID
57489
Gene Name
outer dense fiber of sperm tails 2 like
Gene Aliases
dJ977L11.1
Chromosome Location
Chr. 1: 86346824 - 86396345 on Build GRCh38
UniGene ID
Hs.149360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007022.2 NP_001007023.2 7 967
NM_001184765.1 NP_001171694.1 7 974
NM_001184766.1 NP_001171695.1 7 974
NM_020729.2 NP_065780.2 7 974
XM_005271054.3 XP_005271111.1 7 972
XM_011541815.2 XP_011540117.1 7 972
XM_011541819.2 XP_011540121.1 7 972
XM_011541821.2 XP_011540123.1 7 972
XM_011541823.2 XP_011540125.1 7 972
XM_011541824.2 XP_011540126.1 7 972
XM_017001876.1 XP_016857365.1 7 764
XM_017001877.1 XP_016857366.1 7 764
XM_017001879.1 XP_016857368.1 7 972
GenBank mRNA AB033055.1 - 7 887
AK302792.1 - 7 946
AK303209.1 - 7 971
AK308351.1 - 4 497
BC047368.1 - 7 914 5276959
BC091490.1 - 7 873 30367572

View Full Product Details