Gene Transcripts

Gene Symbol
IGSF9
Entrez Gene ID
57549
Gene Name
immunoglobulin superfamily member 9
Gene Aliases
FP18798, IGSF9A, Nrt1
Chromosome Location
Chr. 1: 159927039 - 159945661 on Build GRCh38
UniGene ID
Hs.591472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135050.1 NP_001128522.1 14 2000
NM_020789.3 NP_065840.2 14 1952
XM_017001884.1 XP_016857373.1 14 2065
GenBank mRNA AB037776.1 - 14 2000
AK056650.1 - 4 1829
AY203932.1 - 3 1305
BC030141.2 - 14 1952 4762143

View Full Product Details