Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303402.1 | NP_001290331.1 | 3 | 259 | |
NM_020839.3 | NP_065890.1 | 4 | 338 | ||
XM_005265346.3 | XP_005265403.1 | 4 | 338 | ||
XM_011533971.1 | XP_011532273.1 | 4 | 450 | ||
XM_017006960.1 | XP_016862449.1 | 4 | 450 | ||
GenBank mRNA | AB040882.2 | - | 4 | 309 | |
AF468833.1 | - | 4 | 298 | ||
AK025513.1 | - | 4 | 308 | ||
AK297349.1 | - | 3 | 229 | ||
AL832926.1 | - | 4 | 292 | ||
BC026353.1 | - | 4 | 308 | 4812745 | |
BC037168.2 | - | 4 | 304 | 4843736 | |
DA215447.1 | - | 4 | 338 |