Gene Transcripts

Gene Symbol
ZFAT
Entrez Gene ID
57623
Gene Name
zinc finger and AT-hook domain containing
Gene Aliases
AITD3, ZFAT1, ZNF406
Chromosome Location
Chr. 8: 134477788 - 134713049 on Build GRCh38
UniGene ID
Hs.446172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029939.3 NP_001025110.2 7 2408
NM_001167583.2 NP_001161055.1 6 2249
NM_001174157.1 NP_001167628.1 5 2140
NM_001174158.1 NP_001167629.1 6 2249
NM_001289394.1 NP_001276323.1 7 2485
NM_020863.3 NP_065914.2 6 2326
NR_110323.1 6 2326
XM_011517203.1 XP_011515505.1 6 2223
XM_011517204.2 XP_011515506.1 5 2046
XM_011517206.1 XP_011515508.1 6 2247
XM_017013716.1 XP_016869205.1 5 2121
GenBank mRNA AB040918.1 - 4 1724
AB167738.1 - 6 2315
AB167739.1 - 7 2474
AB167740.1 - 7 2401
AB167741.1 - 6 2242
AK300133.1 - 7 1935
AK300324.1 - 6 2245
AK300471.1 - 6 2311
AL832066.1 - 6 2326
BC012989.2 - 6 2240
BC025423.2 - 4 1918
BC098405.1 - 6 2249
BC101766.1 - 6 2239 8069272
BC101768.1 - 6 2239 8069274
BC143519.1 - 5 2054 9052028

View Full Product Details