Gene Transcripts

Gene Symbol
SFMBT2
Entrez Gene ID
57713
Gene Name
Scm-like with four mbt domains 2
Gene Aliases
Chromosome Location
Chr. 10: 7158624 - 7411486 on Build GRCh38
UniGene ID
Hs.407983
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018039.1 NP_001018049.1 18 2183
NM_001029880.2 NP_001025051.1 18 2285
XM_005252551.2 XP_005252608.1 18 2066
XM_006717490.2 XP_006717553.1 16 2058
XM_011519611.1 XP_011517913.1 18 2284
XM_017016466.1 XP_016871955.1 18 2186
GenBank mRNA AB046837.1 - 18 2183
BC144327.1 - 13 1753
BC152430.1 - 18 2183 8862546

View Full Product Details