Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021630.5 | NP_067643.3 | 5 | 1123 | |
NM_176871.3 | NP_789847.1 | 4 | 465 | ||
NM_198042.3 | NP_932159.1 | 4 | 465 | ||
GenBank mRNA | AF289563.1 | - | 5 | 722 | |
AK024498.1 | - | 4 | 408 | ||
AK027800.1 | - | 5 | 734 | ||
AK092968.1 | - | 5 | 1123 | ||
AY007729.1 | - | 4 | 489 | ||
AY070438.1 | - | 4 | 489 | ||
AY217349.1 | - | 4 | 450 | ||
BC021556.1 | - | 5 | 604 | 3959372 | |
BC071774.1 | - | 5 | 734 | 4593207 | |
DN997519.1 | - | 5 | 409 |