Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142350.1 | NP_001135822.1 | 8 | 868 | |
NM_024339.3 | NP_077315.2 | 8 | 868 | ||
GenBank mRNA | AK075330.1 | - | 8 | 835 | |
AK314086.1 | - | 8 | 846 | ||
AL832548.1 | - | 7 | 472 | ||
BC003118.1 | - | 9 | 748 | 3537243 | |
BC050674.1 | - | 8 | 807 | 6067702 |