Gene Transcripts

Gene Symbol
NOX5
Entrez Gene ID
79400
Gene Name
NADPH oxidase, EF-hand calcium binding domain 5
Gene Aliases
Chromosome Location
Chr. 15: 68930500 - 69057161 on Build GRCh38
UniGene ID
Hs.657932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184779.1 NP_001171708.1 9 1385
NM_001184780.1 NP_001171709.1 9 1512
NM_024505.3 NP_078781.3 9 1469
NR_033671.2 10 1700
NR_033672.1 10 1616
GenBank mRNA AF317889.1 - 6 900
AF325189.1 - 10 1591
AF325190.1 - 10 1675
AF353088.1 - 9 1364
AF353089.1 - 9 1448
AK026011.1 - 6 954
AK074058.1 - 7 1138
AK074071.1 - 8 4587
AK314689.1 - 9 1448
BC125097.1 - 9 1378 40118286
BC125098.1 - 10 1482 40118288
DQ413001.1 - 6 828

View Full Product Details