Gene Transcripts

Gene Symbol
NKAIN1
Entrez Gene ID
79570
Gene Name
Na+/K+ transporting ATPase interacting 1
Gene Aliases
FAM77C
Chromosome Location
Chr. 1: 31179270 - 31239887 on Build GRCh38
UniGene ID
Hs.470259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024522.2 NP_078798.2 7 1536
XM_017002320.1 XP_016857809.1 6 1130
GenBank mRNA AK022712.1 - 6 1108
AK298874.1 - 5 1028

View Full Product Details