Gene Transcripts
- Gene Symbol
- PARP8
- Entrez Gene ID
- 79668
- Gene Name
- poly(ADP-ribose) polymerase family member 8
- Gene Aliases
- ARTD16, pART16
- Chromosome Location
- Chr. 5: 50665899 - 50846522 on Build GRCh38
- UniGene ID
- Hs.369581
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001178055.1 |
NP_001171526.1
|
16, 17 |
1858 |
|
NM_024615.3 |
NP_078891.2
|
15, 16 |
1834 |
|
XM_005248596.4 |
XP_005248653.1
|
16, 17 |
2214 |
|
XM_011543631.2 |
XP_011541933.1
|
16, 17 |
1747 |
|
XM_011543633.2 |
XP_011541935.2
|
16, 17 |
1794 |
|
XM_011543634.2 |
XP_011541936.1
|
17, 18 |
2343 |
|
XM_011543635.2 |
XP_011541937.1
|
17, 18 |
1864 |
|
XM_011543636.2 |
XP_011541938.1
|
16, 17 |
1728 |
|
XM_011543640.2 |
XP_011541942.1
|
13, 14 |
1618 |
|
XM_011543641.2 |
XP_011541943.1
|
14, 15 |
1595 |
|
XM_011543642.2 |
XP_011541944.1
|
12, 13 |
1582 |
|
XM_011543643.2 |
XP_011541945.1
|
13, 14 |
2023 |
|
XM_017009850.1 |
XP_016865339.1
|
15, 16 |
2294 |
|
XM_017009851.1 |
XP_016865340.1
|
14, 15 |
1734 |
|
GenBank mRNA |
AK024961.1 |
- |
5, 6 |
904 |
|
AK225188.1 |
- |
5, 6 |
904 |
|
AK289369.1 |
- |
15, 16 |
1817 |
|
AK298680.1 |
- |
14, 15 |
1697 |
|
AK314466.1 |
- |
16, 17 |
1858 |
|
AL834477.1 |
- |
15, 16 |
1793 |
|
View Full Product Details