Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024632.5 | NP_078908.1 | 1, 2 | 842 | |
NR_024084.1 | 1, 2 | 842 | |||
GenBank mRNA | AI144341.1 | - | 1, 2 | 254 | |
AK021588.1 | - | 1, 2 | 842 | ||
AY341060.1 | - | 1, 2 | 785 | ||
AY846876.1 | - | 1, 2 | 194 | ||
BC009829.2 | - | 1, 2 | 776 | 3937610 |