Gene Transcripts

Gene Symbol
SEPW1
Entrez Gene ID
6415
Gene Name
selenoprotein W, 1
Gene Aliases
selW
Chromosome Location
Chr. 19: 47778585 - 47784682 on Build GRCh38
UniGene ID
Hs.631549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003009.2 NP_003000.1 4, 5 377
GenBank mRNA AF015283.1 - 4, 5 226
AK026717.1 - 3, 4 1939
BC000581.2 - 4, 5 242 3162433
BC032546.1 - 4, 5 267 5535970
BC039597.1 - 4, 5 245 6015153
BC047893.1 - 4, 5 244 6204763
BF973456.1 - 4, 5 390
U67171.1 - 4, 5 237

View Full Product Details