Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024637.4 | NP_078913.3 | 4 | 1610 | |
GenBank mRNA | AF289575.1 | - | 3 | 1405 | |
AF316113.1 | - | 4 | 1453 | ||
AK022178.1 | - | 3 | 1249 | ||
AK092367.1 | - | 3 | 2236 | ||
AK225890.1 | - | 3 | 1386 | ||
AK301591.1 | - | 2 | 1048 | ||
AL833824.1 | - | 3 | 1255 | ||
BC012976.2 | - | 4 | 1429 | 3636329 |