Gene Transcripts

Gene Symbol
TBC1D17
Entrez Gene ID
79735
Gene Name
TBC1 domain family member 17
Gene Aliases
Chromosome Location
Chr. 19: 49877425 - 49888750 on Build GRCh38
UniGene ID
Hs.631587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168222.1 NP_001161694.1 8, 9 1231
NM_024682.2 NP_078958.2 9, 10 1330
XM_011527317.2 XP_011525619.1 9, 10 1242
GenBank mRNA AB449903.1 - 9, 10 1031
AK022230.1 - 9, 10 1089
AK090606.1 - 8, 9 1031
AK300007.1 - 8, 9 990
AY129013.1 - 7, 8 1777
BC003516.1 - 9, 10 1183 3050014

View Full Product Details