Gene Transcripts

Gene Symbol
MMRN2
Entrez Gene ID
79812
Gene Name
multimerin 2
Gene Aliases
EMILIN-3, EMILIN3, ENDOGLYX-1
Chromosome Location
Chr. 10: 86935540 - 86971311 on Build GRCh38
UniGene ID
Hs.524479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024756.2 NP_079032.2 2 336
XM_005270153.1 XP_005270210.1 2 375
XM_006717970.3 XP_006718033.1 2 2315
GenBank mRNA AK023527.1 - 2 283
AK293822.1 - 2 283
AK304727.1 - 2 193
BC064415.1 - 2 291
BC072456.1 - 2 232
BC094744.1 - 2 283 30343159
DC416005.1 - 2 337

View Full Product Details