Gene Transcripts
- Gene Symbol
- PLEKHS1
- Entrez Gene ID
- 79949
- Gene Name
- pleckstrin homology domain containing S1
- Gene Aliases
- C10orf81, HEL185, bA211N11.2
- Chromosome Location
- Chr. 10: 113751262 - 113782433 on Build GRCh38
- UniGene ID
- Hs.228320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001193434.1 |
NP_001180363.1
|
8, 9 |
1099 |
|
NM_001193435.1 |
NP_001180364.1
|
6, 7 |
903 |
|
NM_024889.4 |
NP_079165.3
|
8, 9 |
999 |
|
NM_182601.1 |
NP_872407.1
|
7, 8 |
1216 |
|
XM_005270162.2 |
XP_005270219.1
|
8, 9 |
1138 |
|
XM_005270163.2 |
XP_005270220.1
|
8, 9 |
1138 |
|
XM_011540171.1 |
XP_011538473.1
|
8, 9 |
1138 |
|
XM_011540172.1 |
XP_011538474.1
|
9, 10 |
906 |
|
XM_011540173.1 |
XP_011538475.1
|
8, 9 |
1138 |
|
XM_011540174.2 |
XP_011538476.1
|
8, 9 |
1138 |
|
XM_011540175.1 |
XP_011538477.1
|
8, 9 |
1077 |
|
XM_011540176.1 |
XP_011538478.1
|
6, 7 |
675 |
|
XM_017016666.1 |
XP_016872155.1
|
8, 9 |
1138 |
|
GenBank mRNA |
AK027190.1 |
- |
1, 2 |
62 |
|
AK126354.1 |
- |
6, 7 |
675 |
|
AK128811.1 |
- |
8, 9 |
999 |
|
AK292867.1 |
- |
8, 9 |
871 |
|
BC036365.1 |
- |
2, 3 |
153 |
|
BC037870.1 |
- |
7, 8 |
1221 |
5273287 |
FJ236306.1 |
- |
7, 8 |
1221 |
|
View Full Product Details