Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004388.2 | NP_004379.1 | 6 | 1009 | |
GenBank mRNA | AK075423.1 | - | 6 | 1273 | |
AK298703.1 | - | 6 | 960 | ||
AK314640.1 | - | 6 | 975 | ||
BC024007.2 | - | 5 | 791 | ||
BC096752.1 | - | 5 | 791 | ||
BC126333.1 | - | 6 | 985 | 8992049 | |
BC126335.1 | - | 6 | 985 | 8992051 | |
M95767.1 | - | 6 | 944 |