Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253875.1 | NP_001240804.1 | 7 | 655 | |
NM_001253876.1 | NP_001240805.1 | 2 | 792 | ||
NM_025076.4 | NP_079352.2 | 7 | 640 | ||
NR_045607.1 | 6 | 591 | |||
XM_006712775.3 | XP_006712838.1 | 7 | 617 | ||
XM_011511902.2 | XP_011510204.2 | 7 | 634 | ||
XM_011511903.2 | XP_011510205.2 | 7 | 617 | ||
XM_011511904.2 | XP_011510206.1 | 4 | 962 | ||
XM_017005008.1 | XP_016860497.1 | 7 | 617 | ||
XM_017005009.1 | XP_016860498.1 | 7 | 602 | ||
XM_017005010.1 | XP_016860499.1 | 7 | 602 | ||
XM_017005011.1 | XP_016860500.1 | 7 | 742 | ||
XM_017005012.1 | XP_016860501.1 | 7 | 677 | ||
XM_017005013.1 | XP_016860502.1 | 5 | 1200 | ||
XM_017005014.1 | XP_016860503.1 | 6 | 418 | ||
GenBank mRNA | AK027244.1 | - | 2 | 792 | |
AK075120.1 | - | 7 | 602 | ||
AK075170.1 | - | 7 | 617 | ||
AK225592.1 | - | 2 | 792 | ||
AK290668.1 | - | 7 | 615 | ||
AY147934.1 | - | 7 | 607 | ||
AY358541.1 | - | 7 | 591 | ||
BC009819.2 | - | 7 | 581 | 3843312 | |
CF242897.1 | - | 7 | 581 |