Gene Transcripts
- Gene Symbol
- CWH43
- Entrez Gene ID
- 80157
- Gene Name
- cell wall biogenesis 43 C-terminal homolog
- Gene Aliases
- CWH43-C
- Chromosome Location
- Chr. 4: 48986247 - 49062079 on Build GRCh38
- UniGene ID
- Hs.479703
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286791.1 |
NP_001273720.1
|
14, 15 |
1965 |
|
NM_025087.2 |
NP_079363.2
|
14, 15 |
2046 |
|
XM_011513755.1 |
XP_011512057.1
|
14, 15 |
2104 |
|
XM_011513756.2 |
XP_011512058.1
|
14, 15 |
2135 |
|
XM_011513757.2 |
XP_011512059.1
|
14, 15 |
2161 |
|
XM_011513758.1 |
XP_011512060.1
|
13, 14 |
1846 |
|
XM_011513759.1 |
XP_011512061.1
|
11, 12 |
1500 |
|
GenBank mRNA |
AK025164.1 |
- |
14, 15 |
2046 |
|
AK300495.1 |
- |
14, 15 |
1968 |
|
AK316307.1 |
- |
14, 15 |
1938 |
|
BC137387.1 |
- |
14, 15 |
2029 |
9021385 |
BC137388.1 |
- |
14, 15 |
2029 |
9021386 |
View Full Product Details