Gene Transcripts

Gene Symbol
CTBP2
Entrez Gene ID
1488
Gene Name
C-terminal binding protein 2
Gene Aliases
Chromosome Location
Chr. 10: 124987849 - 125161061 on Build GRCh38
UniGene ID
Hs.501345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083914.2 NP_001077383.1 11 2234
NM_001290214.2 NP_001277143.1 11 2219
NM_001290215.2 NP_001277144.1 11 2483
NM_001321012.1 NP_001307941.1 11 2178
NM_001321013.1 NP_001307942.1 10 2379
NM_001321014.1 NP_001307943.1 11 2480
NM_001329.3 NP_001320.1 11 2222
NM_022802.2 NP_073713.2 9 3539
XM_005269561.2 XP_005269618.1 12 2405
XM_005269564.2 XP_005269621.1 12 2613
XM_005269567.2 XP_005269624.1 12 2612
XM_005269568.4 XP_005269625.1 12 2340
XM_005269569.2 XP_005269626.1 11 2301
XM_005269571.2 XP_005269628.1 10 2060
XM_005269572.3 XP_005269629.1 10 2108
XM_006717642.2 XP_006717705.1 13 2534
XM_011539349.2 XP_011537651.1 9 2316
XM_011539351.1 XP_011537653.1 14 2635
XM_011539353.1 XP_011537655.1 13 2506
XM_011539354.1 XP_011537656.1 12 2430
XM_011539355.1 XP_011537657.1 11 2508
XM_011539357.1 XP_011537659.1 11 2246
XM_011539358.1 XP_011537660.1 10 2324
XM_017015756.1 XP_016871245.1 12 2276
XM_017015757.1 XP_016871246.1 11 4002
GenBank mRNA AF016507.1 - 11 2139
AF222711.1 - 9 3557
AL833398.1 - 11 2194
BC052276.1 - 11 2176

View Full Product Details