Gene Transcripts
- Gene Symbol
- FHOD3
- Entrez Gene ID
- 80206
- Gene Name
- formin homology 2 domain containing 3
- Gene Aliases
- FHOS2, Formactin2
- Chromosome Location
- Chr. 18: 36297696 - 36780220 on Build GRCh38
- UniGene ID
- Hs.436636
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281740.2 |
NP_001268669.1
|
15, 16 |
2119 |
|
NM_025135.4 |
NP_079411.2
|
12, 13 |
1594 |
|
XM_005258349.1 |
XP_005258406.1
|
13, 14 |
1912 |
|
XM_005258354.1 |
XP_005258411.1
|
11, 12 |
1462 |
|
XM_005258355.1 |
XP_005258412.1
|
11, 12 |
1462 |
|
XM_011526189.1 |
XP_011524491.1
|
16, 17 |
2161 |
|
XM_011526190.1 |
XP_011524492.1
|
16, 17 |
2161 |
|
XM_011526192.1 |
XP_011524494.1
|
15, 16 |
2086 |
|
XM_011526193.2 |
XP_011524495.1
|
16, 17 |
2161 |
|
XM_011526195.1 |
XP_011524497.1
|
16, 17 |
2161 |
|
XM_011526196.1 |
XP_011524498.1
|
13, 14 |
1636 |
|
XM_011526197.1 |
XP_011524499.1
|
15, 16 |
2086 |
|
XM_017026006.1 |
XP_016881495.1
|
16, 17 |
2161 |
|
XM_017026007.1 |
XP_016881496.1
|
14, 15 |
1799 |
|
XM_017026008.1 |
XP_016881497.1
|
16, 17 |
2161 |
|
XM_017026009.1 |
XP_016881498.1
|
12, 13 |
1576 |
|
XM_017026010.1 |
XP_016881499.1
|
14, 15 |
1805 |
|
GenBank mRNA |
AB051482.1 |
- |
5, 6 |
735 |
|
BC031518.1 |
- |
8, 9 |
1046 |
|
HM191478.1 |
- |
15, 16 |
1979 |
|
View Full Product Details