Gene Transcripts

Gene Symbol
OR10W1
Entrez Gene ID
81341
Gene Name
olfactory receptor family 10 subfamily W member 1
Gene Aliases
OR10W1P, OR10W1Q, OR11-236, UNQ6469
Chromosome Location
Chr. 11: 58266792 - 58268260 on Build GRCh38
UniGene ID
Hs.531507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207374.3 NP_997257.2 1 1242
GenBank mRNA AY358251.1 - 1 1242
BC132848.1 - 1 1141 40146870
BC132850.1 - 1 1141 40146872

View Full Product Details