Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000397.3 | NP_000388.2 | 12, 13 | 1650 | |
GenBank mRNA | AK289753.1 | - | 12, 13 | 1635 | |
AK301476.1 | - | 10, 11 | 1257 | ||
AK312929.1 | - | 12, 13 | 1611 | ||
BC032720.1 | - | 12, 13 | 1636 | 5505170 | |
KU178009.1 | - | 12, 13 | 1589 | ||
KU178010.1 | - | 6, 7 | 562 | ||
X04011.1 | - | 12, 13 | 1604 |